missing translation for 'onlineSavingsMsg'
Learn More

Ofloxacin 98.0+%, TCI America™

Catalog No. p-7134318
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O04035G 5 g
O040325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O04035G Supplier TCI America Supplier No. O04035G

Chemical Identifiers

CAS 82419-36-1
Molecular Formula C18H20FN3O4
Molecular Weight (g/mol) 361.373
MDL Number MFCD00226105
InChI Key GSDSWSVVBLHKDQ-UHFFFAOYSA-N
Synonym ofloxacin, ofloxacine, floxin, tarivid, oxaldin, ofloxacino, ofloxacinum, ocuflox, oflocet, visiren
PubChem CID 4583
ChEBI CHEBI:7731
SMILES CC1COC2=C3N1C=C(C(=O)C3=CC(=C2N4CCN(CC4)C)F)C(=O)O

Specifications

Melting Point 263°C
Color White-Yellow
Quantity 5 g
Formula Weight 361.37
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ofloxacin
RTECSNumber UU8815500
TSCA No