missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ofloxacin 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 263°C |
| Color | White-Yellow |
| Quantity | 25 g |
| Formula Weight | 361.37 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Ofloxacin |
Safety and Handling
| RTECSNumber | UU8815500 |
| TSCA | No |