missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Office Upper Flipper Door Cabinet

Catalog No. 06001044
Change view
Click to view available options
Dimensions (D x W x H):
13 x 30 x 15 in.
13 x 36 x 15 in.
13 x 48 x 15 in.
13 x 60 x 15 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (D x W x H)
06001044 13 x 30 x 15 in.
06001045 13 x 36 x 15 in.
06001046 13 x 48 x 15 in.
06001047 13 x 60 x 15 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 06001044 Supplier Fisherbrand™ Supplier No. FSPFDU30DT

Upper flipper door cabinets are used for added storage space on 67 in. tall systems without glass panels.

  • Flipper door cabinet with lock
  • Dark tone color
  • Used on systems without glass panels

Specifications

Dimensions (D x W x H) 13 x 30 x 15 in.
For Use With (Application) Office Workstations