missing translation for 'onlineSavingsMsg'
Learn More

ODQ, 98%

Catalog No. AC328800100
Change view
Click to view available options
Quantity:
10 mg
50 mg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC328800100 10 mg Glass bottle
AC328800500 50 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC328800100 Supplier Thermo Scientific Chemicals Supplier No. 328800100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 41443-28-1 | C9H5N3O2 | 187.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 41443-28-1
Molecular Formula C9H5N3O2
Molecular Weight (g/mol) 187.2
MDL Number MFCD00792620
InChI Key LZMHWZHOZLVYDL-UHFFFAOYSA-N
Synonym 1h-1,2,4 oxadiazolo 4,3-a quinoxalin-1-one, odq, 1,2,4 oxadiazolo 4,3-a quinoxalin-1-one, unii-s57v2nmv38, 1h-1,2,4 oxadiazolo 4,3-a-quinoxalin-1-one, 1h-1,2,4 oxadiazolo-4,3,a quinoxalin-1-one, 1-h-1,2,4 oxadiazolo-4,3,2 quinoxalin-1-ine, 1h-odq
PubChem CID 1456
IUPAC Name [1,2,4]oxadiazolo[4,3-a]quinoxalin-1-one
SMILES C1=CC=C2C(=C1)N=CC3=NOC(=O)N23

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 160°C to 170°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 10 mg
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in chloroform,ethylacetate,dmso
Formula Weight 187.2
Percent Purity 98%
Physical Form Powder
Chemical Name or Material ODQ
Show More Show Less

RUO – Research Use Only