missing translation for 'onlineSavingsMsg'
Learn More

Decon™ OdorGone™ Autoclave Deodorizer
GREENER_CHOICE

Catalog No. 22032080
Change view
Click to view available options
Scent:
Apple
Lemon
Quantity:
100 Capsules
25 Capsules
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Scent Quantity
22032080 Apple 100 Capsules
22032085 Lemon 25 Capsules
22032087 Lemon 100 Capsules
22032079 Apple 25 Capsules
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 22032080 Supplier Decon™ Supplier No. APP100
Edge

missing translation for 'validMainframeMsg'

High concentration liquid deodorant in capsule form

One capsule placed in each load absorbs offensive odors, leaving your autoclaves smelling fresh and clean.

Order Info

Available in Apple and Lemon fragrances

Specifications

Product Type Autoclave Capsule
Color-coded No
Form Liquid
Scent Apple
Sterility Non-sterile
Autoclavable Autoclavable
Color Translucent Red
Format Capsules
For Use With (Application) Autoclave Deodorizer
Quantity 100 Capsules