missing translation for 'onlineSavingsMsg'
Learn More

Octyl sulfate, sodium salt, 99%, HPLC grade

Catalog No. AC416501000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC416501000 100 g Glass bottle
AC416500250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC416501000 Supplier Thermo Scientific Chemicals Supplier No. 416501000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 142-31-4 | C8H15Na·H2SO4 | 232.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ion pair chromatography

Chemical Identifiers

CAS 142-31-4
Molecular Formula C8H15Na·H2SO4
Molecular Weight (g/mol) 232.28
InChI Key WFRKJMRGXGWHBM-UHFFFAOYSA-M
Synonym sodium octyl sulfate, sodium n-octyl sulfate, sodium octyl sulphate, sipex ols, cycloryl os, duponol 80, sodium capryl sulfate, octyl sodium sulfate, sulfuric acid, monooctyl ester, sodium salt, octyl sulfate sodium salt
PubChem CID 2735107
IUPAC Name sodium;octyl sulfate
SMILES CCCCCCCCOS(=O)(=O)[O-].[Na+]

Specifications

Absorbance (5% aq. soln.) (1cm cell), (5% aq. soln.) (1 cm cell) at 0nm, 0.02 max. at 250nm
Melting Point 210.0°C
Color White
Loss on Drying 2% (105°C) (vacuum/P2O5) max.
Infrared Spectrum Authentic
Assay Percent Range 1% max. Alkyl sulphate homologues (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)7OSO3Na
Quantity 100 g
Solubility Information (25% in water at 25°C) Clear colorless
Formula Weight 232.28
Percent Purity 99%
Physical Form Coarse Powder
Chemical Name or Material Octyl sulfate, sodium salt
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye damage.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
EINECSNumber 205-535-5

RUO – Research Use Only