missing translation for 'onlineSavingsMsg'
Learn More

Octyl Phenol Ethoxylate, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. 02003563
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Narrow Mouth Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02003563 500 mL Narrow Mouth Amber Glass Bottle
02003561 120 mL Narrow Mouth Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02003563 Supplier Avantor Supplier No. X19807

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High quality chemicals for laboratory and specialized industrial use

Specifications

Flash Point 251.00°C
Packaging Narrow Mouth Amber Glass Bottle
Quantity 500 mL
Specific Gravity 1L = 1.07kg
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Octyl Phenol Ethoxylate