missing translation for 'onlineSavingsMsg'
Learn More

Octyl aldehyde, 99%

Catalog No. p-177037
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC129481000 100 mL Glass bottle
AC129485000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC129481000 Supplier Thermo Scientific Chemicals Supplier No. 129481000
Only null left

CAS: 124-13-0 | C8H16O | 128.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 124-13-0
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.22
MDL Number MFCD00007029
InChI Key NUJGJRNETVAIRJ-UHFFFAOYSA-N
Synonym caprylic aldehyde, caprylaldehyde, n-octanal, 1-octanal, n-octyl aldehyde, n-octaldehyde, n-caprylaldehyde, octanaldehyde, aldehyde c-8, n-octylal
PubChem CID 454
ChEBI CHEBI:17935
IUPAC Name octanal
SMILES CCCCCCCC=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 12.0°C to 15.0°C
Color Colorless to Yellow
Density 0.8210g/mL
Boiling Point 171.0°C
Flash Point 51°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4170 to 1.4200
Linear Formula CH3(CH2)6CHO
Quantity 100 mL
Beilstein 01, 704
Specific Gravity 0.821
Merck Index 15, 1768
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and mineral oil, insoluble in glycerol
Formula Weight 128.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Octyl aldehyde, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta
EINECSNumber 204-683-8
RTECSNumber RG7780000
TSCA TSCA

RUO – Research Use Only