missing translation for 'onlineSavingsMsg'
Learn More

Octanenitrile 97.0+%, TCI America™

Catalog No. p-7134243
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O022125ML 25 mL
O0221250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O022125ML Supplier TCI America Supplier No. O022125ML

Chemical Identifiers

CAS 124-12-9
Molecular Formula C8H15N
Molecular Weight (g/mol) 125.22
MDL Number MFCD00001983
InChI Key YSIMAPNUZAVQER-UHFFFAOYSA-N
Synonym caprylonitrile, octanonitrile, arneel 8, 1-cyanoheptane, caprylnitrile, n-heptyl cyanide, heptyl cyanide, heptylcyanide, unii-d6646v87py, n-heptylcyanide
PubChem CID 31286
IUPAC Name octanenitrile
SMILES CCCCCCCC#N

Specifications

Melting Point -45°C
Color Yellow
Boiling Point 200°C
Quantity 25 mL
Formula Weight 125.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Octanenitrile
EINECSNumber (2)-1507
RTECSNumber RG9700300
TSCA Yes