missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octamethyltrisiloxane 98.0+%, TCI America™
$102.52 - $262.83
Chemical Identifiers
| CAS | 107-51-7 |
|---|---|
| Molecular Formula | C8H24O2Si3 |
| Molecular Weight (g/mol) | 236.53 |
| MDL Number | MFCD00084411,MFCD00134211,MFCD00148360,MFCD00008264,MFCD00132673 |
| InChI Key | CXQXSVUQTKDNFP-UHFFFAOYSA-N |
| Synonym | Dimethylbis(trimethylsilyloxy)silane |
| PubChem CID | 24705 |
| ChEBI | CHEBI:9147 |
| IUPAC Name | 2,2,4,4,6,6-hexamethyl-3,5-dioxa-2,4,6-trisilaheptane |
| SMILES | C[Si](C)(C)O[Si](C)(C)O[Si](C)(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
O025725ML
|
TCI America
O025725ML |
25 mL |
Each for $102.52
|
|
|||||
|
O0257100ML
|
TCI America
O0257100ML |
100 mL |
Each for $262.83
|
|
|||||
Chemical Identifiers
| 107-51-7 | |
| 236.53 | |
| CXQXSVUQTKDNFP-UHFFFAOYSA-N | |
| 24705 | |
| 2,2,4,4,6,6-hexamethyl-3,5-dioxa-2,4,6-trisilaheptane |
| C8H24O2Si3 | |
| MFCD00084411,MFCD00134211,MFCD00148360,MFCD00008264,MFCD00132673 | |
| Dimethylbis(trimethylsilyloxy)silane | |
| CHEBI:9147 | |
| C[Si](C)(C)O[Si](C)(C)O[Si](C)(C)C |
Specifications
| 107-51-7 | |
| 153°C | |
| MFCD00084411,MFCD00134211,MFCD00148360,MFCD00008264,MFCD00132673 | |
| 1993 | |
| CXQXSVUQTKDNFP-UHFFFAOYSA-N | |
| 2,2,4,4,6,6-hexamethyl-3,5-dioxa-2,4,6-trisilaheptane | |
| 24705 | |
| 236.53 | |
| Liquid |
| Colorless | |
| C8H24O2Si3 | |
| 25 mL | |
| Dimethylbis(trimethylsilyloxy)silane | |
| C[Si](C)(C)O[Si](C)(C)O[Si](C)(C)C | |
| 236.53 | |
| CHEBI:9147 | |
| ≥98.0% (GC) | |
| Octamethyltrisiloxane |
Safety and Handling
EINECSNumber : (7)-0476
TSCA : Yes