Learn More
Octamethylcyclotetrasiloxane, 98%
Description
Octamethylcyclotetrasiloxane is used to prepare silicone oil, silicone rubber and silicone resin having different degrees of polymerization through ring opening and polymerizing. It is also widely used in various fields viz. construction, electronics, textile, automobile, personal care, food, and machining. It is an organometallic reagent utilized for the preparation of siloxane polymers having high surface-hydrophobicity restoration.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Octamethylcyclotetrasiloxane is used to prepare silicone oil, silicone rubber and silicone resin having different degrees of polymerization through ring opening and polymerizing. It is also widely used in various fields viz. construction, electronics, textile, automobile, personal care, food, and machining. It is an organometallic reagent utilized for the preparation of siloxane polymers having high surface-hydrophobicity restoration.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents, acids and bases.
Specifications
Specifications
| Boiling Point | 175°C to 176°C |
| Melting Point | 17°C to 18°C |
| Quantity | 500 g |
| Flash Point | 56°C (132°F) |
| Refractive Index | 1.396 |
| Sensitivity | Moisture sensitive |
| Density | 0.956 |
| UN Number | UN1993 |
| Beilstein | 1787074 |
| Merck Index | 14,6747 |
| Show More |
Safety and Handling
| Hazard Category | H226-H302-H312-H360 |
| Hazard Statement | GHS H Statement H226-H361 Flammable liquid and vapour. Suspected of damaging fertility or the unborn child. |
| Precautionary Statement | P201-P202-P210-P233-P240-P241-P242-P243-P264b-P270-P281-P301+P312-P303+P361+P353-P308+P313-P312-P330-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
| EINECSNumber | 209-136-7 |
| RTECSNumber | GZ4397000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only