missing translation for 'onlineSavingsMsg'
Learn More

Octamethylcyclotetrasiloxane, 98%

Catalog No. AAA1524022
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1524022 100 g
AAA1524036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1524022 Supplier Thermo Scientific Chemicals Supplier No. A1524022

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 556-67-2 | C8H24O4Si4 | 296.616 g/mol

Octamethylcyclotetrasiloxane is used to prepare silicone oil, silicone rubber and silicone resin having different degrees of polymerization through ring opening and polymerizing. It is also widely used in various fields viz. construction, electronics, textile, automobile, personal care, food, and machining. It is an organometallic reagent utilized for the preparation of siloxane polymers having high surface-hydrophobicity restoration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Octamethylcyclotetrasiloxane is used to prepare silicone oil, silicone rubber and silicone resin having different degrees of polymerization through ring opening and polymerizing. It is also widely used in various fields viz. construction, electronics, textile, automobile, personal care, food, and machining. It is an organometallic reagent utilized for the preparation of siloxane polymers having high surface-hydrophobicity restoration.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents, acids and bases.

Chemical Identifiers

CAS 556-67-2
Molecular Formula C8H24O4Si4
Molecular Weight (g/mol) 296.616
MDL Number MFCD00003269
InChI Key HMMGMWAXVFQUOA-UHFFFAOYSA-N
Synonym octamethylcyclotetrasiloxane, cyclotetrasiloxane, octamethyl, oktamethylcyklotetrasiloxan, cyclic dimethylsiloxane tetramer, omcts, nuc silicone vs 7207, union carbide 7207, silicone sf 1173, oktamethylzyklotetrasiloxan, unii-cz227117je
PubChem CID 11169
ChEBI CHEBI:25640
IUPAC Name 2,2,4,4,6,6,8,8-octamethyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane
SMILES C[Si]1(O[Si](O[Si](O[Si](O1)(C)C)(C)C)(C)C)C

Specifications

Boiling Point 175°C to 176°C
Melting Point 17°C to 18°C
Quantity 100 g
Flash Point 56°C (132°F)
Refractive Index 1.396
Sensitivity Moisture sensitive
Density 0.956
UN Number UN1993
Beilstein 1787074
Merck Index 14,6747
Solubility Information Immiscible with water.
Formula Weight 296.62
Percent Purity 98%
Chemical Name or Material Octamethylcyclotetrasiloxane
Show More Show Less
Hazard Category H226-H302-H312-H360
Hazard Statement GHS H Statement
H226-H361
Flammable liquid and vapour.
Suspected of damaging fertility or the unborn child.
Precautionary Statement P201-P202-P210-P233-P240-P241-P242-P243-P264b-P270-P281-P301+P312-P303+P361+P353-P308+P313-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 209-136-7
RTECSNumber GZ4397000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only