missing translation for 'onlineSavingsMsg'
Learn More

Octamethylcyclotetrasilazane 96.0+%, TCI America™

Catalog No. O015310G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O015310G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O015310G Supplier TCI America Supplier No. O015310G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1020-84-4
Molecular Formula C8H28N4Si4
Molecular Weight (g/mol) 292.68
MDL Number MFCD00046123
InChI Key FIADVASZMLCQIF-UHFFFAOYSA-N
Synonym Dimethylsilazane Cyclic Tetramer
IUPAC Name 2,2,4,4,6,6,8,8-octamethyl-1,3,5,7,2,4,6,8-tetrazatetrasilocane
SMILES C[Si]1(C)N[Si](C)(C)N[Si](C)(C)N[Si](C)(C)N1

Specifications

Boiling Point 225°C
Melting Point 98°C
Color White
Physical Form Crystalline Powder
Quantity 10 g
UN Number 2923
Formula Weight 292.68
Percent Purity ≥96.0% (T)
Chemical Name or Material Octamethylcyclotetrasilazane
RTECSNumber GZ4342500
TSCA Yes