missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octamethylcyclotetrasilazane, 97%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 224°C to 225°C |
| Melting Point | 94°C to 98°C |
| Quantity | 5 g |
| UN Number | UN2923 |
| Beilstein | 976248 |
| Flash Point | 66°C (150°F) |
| Formula Weight | 292.68 |
| Percent Purity | 97% |
| Odor | Ammonia-like |
| Sensitivity | Moisture sensitive |
| Show More |
Safety and Handling
| Hazard Category | H301-H314 |
| Hazard Statement | GHS H Statement H301-H314-H318 Toxic if swallowed. Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S. |
| EINECSNumber | 213-817-4 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only