missing translation for 'onlineSavingsMsg'
Learn More

Octamethylcyclotetrasilazane, 97%

Catalog No. AAL1656306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1656306 5 g
AAL1656314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1656306 Supplier Thermo Scientific Chemicals Supplier No. L1656306
Only null left

CAS: 1020-84-4 | C8H28N4Si4 | 292.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1020-84-4
Molecular Formula C8H28N4Si4
Molecular Weight (g/mol) 292.68
MDL Number MFCD00046123
InChI Key FIADVASZMLCQIF-UHFFFAOYSA-N
IUPAC Name 2,2,4,4,6,6,8,8-octamethyl-1,3,5,7,2,4,6,8-tetrazatetrasilocane
SMILES C[Si]1(C)N[Si](C)(C)N[Si](C)(C)N[Si](C)(C)N1

Specifications

Boiling Point 224°C to 225°C
Melting Point 94°C to 98°C
Quantity 5 g
UN Number UN2923
Beilstein 976248
Flash Point 66°C (150°F)
Formula Weight 292.68
Percent Purity 97%
Odor Ammonia-like
Sensitivity Moisture sensitive
Chemical Name or Material Octamethylcyclotetrasilazane
Show More Show Less
Hazard Category H301-H314
Hazard Statement GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber 213-817-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only