missing translation for 'onlineSavingsMsg'
Learn More

Octafluorocyclopentene 98.0+%, TCI America™

Catalog No. p-7134281
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O029210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O029210G Supplier TCI America Supplier No. O029210G

Chemical Identifiers

CAS 559-40-0
Molecular Formula C5F8
Molecular Weight (g/mol) 212.042
MDL Number MFCD00013745
InChI Key YBMDPYAEZDJWNY-UHFFFAOYSA-N
Synonym Perfluorocyclopentene
PubChem CID 11212
IUPAC Name 1,2,3,3,4,4,5,5-octafluorocyclopentene
SMILES C1(=C(C(C(C1(F)F)(F)F)(F)F)F)F

Specifications

Color Colorless
Quantity 10 g
Formula Weight 212.04
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Octafluorocyclopentene
EINECSNumber (3)-4350
RTECSNumber GY6000000
TSCA Yes
Recommended Storage Refrigerator