missing translation for 'onlineSavingsMsg'
Learn More

Octaethylene Glycol 96.0+%, TCI America™

Catalog No. p-7134283
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02951G 1 g
O02955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O02951G Supplier TCI America Supplier No. O02951G
Only null left

Chemical Identifiers

CAS 5117-19-1
Molecular Formula C16H34O9
Molecular Weight (g/mol) 370.439
MDL Number MFCD00698694
InChI Key GLZWNFNQMJAZGY-UHFFFAOYSA-N
PubChem CID 78798
ChEBI CHEBI:44794
IUPAC Name 2-[2-[2-[2-[2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethanol
SMILES C(COCCOCCOCCOCCOCCOCCOCCO)O

Specifications

Color Yellow
Quantity 1 g
Formula Weight 370.44
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Octaethylene Glycol
EINECSNumber (2)-0441
TSCA No