missing translation for 'onlineSavingsMsg'
Learn More

Octadecyltriethoxysilane 85.0+%, TCI America™

Catalog No. O016525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O016525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O016525G Supplier TCI America Supplier No. O016525G
Only null left

Chemical Identifiers

CAS 7399-00-0
Molecular Formula C24H52O3Si
Molecular Weight (g/mol) 416.76
MDL Number MFCD00039875
InChI Key FZMJEGJVKFTGMU-UHFFFAOYSA-N
Synonym octadecyltriethoxysilane, triethoxy octadecyl silane, n-octadecyltriethoxysilane, octadecyl triethoxysilane, stearyl triethoxysilane, unii-1vn9p25l8h, triethoxystearylsilane, acmc-209ot8
PubChem CID 81885
IUPAC Name triethoxy(octadecyl)silane
SMILES CCCCCCCCCCCCCCCCCC[Si](OCC)(OCC)OCC

Specifications

Melting Point 12°C
Color Yellow
Boiling Point 169°C
Quantity 25 g
Formula Weight 416.76
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material Octadecyltriethoxysilane