missing translation for 'onlineSavingsMsg'
Learn More

Octadecyltriethoxysilane, 98%, n-isomer 85% min

CAS: 7399-00-0 | C24H52O3Si | 416.76 g/mol

$88.79 - $300.10

Chemical Identifiers

CAS 7399-00-0
Molecular Formula C24H52O3Si
Molecular Weight (g/mol) 416.76
MDL Number MFCD00039875
InChI Key FZMJEGJVKFTGMU-UHFFFAOYSA-N
Synonym octadecyltriethoxysilane, triethoxy octadecyl silane, n-octadecyltriethoxysilane, octadecyl triethoxysilane, stearyl triethoxysilane, unii-1vn9p25l8h, triethoxystearylsilane, acmc-209ot8
PubChem CID 81885
IUPAC Name triethoxy(octadecyl)silane
SMILES CCCCCCCCCCCCCCCCCC[Si](OCC)(OCC)OCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0636609
View Documents
Thermo Scientific Chemicals
L0636609
10 g
Each for $88.79
Only null left
 
AAL0636618
View Documents
Thermo Scientific Chemicals
L0636618
50 g
Each for $300.10
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7399-00-0
416.76
FZMJEGJVKFTGMU-UHFFFAOYSA-N
81885
CCCCCCCCCCCCCCCCCC[Si](OCC)(OCC)OCC
C24H52O3Si
MFCD00039875
octadecyltriethoxysilane, triethoxy octadecyl silane, n-octadecyltriethoxysilane, octadecyl triethoxysilane, stearyl triethoxysilane, unii-1vn9p25l8h, triethoxystearylsilane, acmc-209ot8
triethoxy(octadecyl)silane

Specifications

165°C to 169°C (0.2mmHg)
10 g
MFCD00039875
195°C (383°F)
FZMJEGJVKFTGMU-UHFFFAOYSA-N
triethoxy(octadecyl)silane
81885
98%
Moisture sensitive
Octadecyltriethoxysilane
7399-00-0
C24H52O3Si
7025163
octadecyltriethoxysilane, triethoxy octadecyl silane, n-octadecyltriethoxysilane, octadecyl triethoxysilane, stearyl triethoxysilane, unii-1vn9p25l8h, triethoxystearylsilane, acmc-209ot8
CCCCCCCCCCCCCCCCCC[Si](OCC)(OCC)OCC
416.76
416.77
1.438
0.87
n-isomer ≥85%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 230-995-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only