missing translation for 'onlineSavingsMsg'
Learn More

Octadecyltriethoxysilane, 98%, n-isomer 85% min

Catalog No. AAL0636618
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0636618 50 g
AAL0636609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0636618 Supplier Thermo Scientific Chemicals Supplier No. L0636618
Only null left

CAS: 7399-00-0 | C24H52O3Si | 416.76 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7399-00-0
Molecular Formula C24H52O3Si
Molecular Weight (g/mol) 416.76
MDL Number MFCD00039875
InChI Key FZMJEGJVKFTGMU-UHFFFAOYSA-N
Synonym octadecyltriethoxysilane, triethoxy octadecyl silane, n-octadecyltriethoxysilane, octadecyl triethoxysilane, stearyl triethoxysilane, unii-1vn9p25l8h, triethoxystearylsilane, acmc-209ot8
PubChem CID 81885
IUPAC Name triethoxy(octadecyl)silane
SMILES CCCCCCCCCCCCCCCCCC[Si](OCC)(OCC)OCC

Specifications

Boiling Point 165°C to 169°C (0.2mmHg)
Quantity 50 g
Beilstein 7025163
Flash Point 195°C (383°F)
Formula Weight 416.77
Percent Purity 98%
Refractive Index 1.438
Sensitivity Moisture sensitive
Density 0.87
Assay n-isomer ≥85%
Chemical Name or Material Octadecyltriethoxysilane
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 230-995-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only