missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octadecylbenzene 95.0+%, TCI America™
$264.70 - $708.33
Chemical Identifiers
| CAS | 4445-07-2 |
|---|---|
| Molecular Formula | C24H42 |
| Molecular Weight (g/mol) | 330.60 |
| MDL Number | MFCD00048500 |
| InChI Key | WSVDSBZMYJJMSB-UHFFFAOYSA-N |
| Synonym | 1-phenyloctadecane, benzene, octadecyl, n-octadecylbenzene, stearylbenzene, 2-phenyloctadecane, octadecane, 1-phenyl, n-octadecyl benzene, acmc-2097fm, octadecane, 1-phenyl-8ci, 1-phenyloctadecane, analytical standard |
| PubChem CID | 78187 |
| IUPAC Name | octadecylbenzene |
| SMILES | CCCCCCCCCCCCCCCCCCC1=CC=CC=C1 |
Chemical Identifiers
| 4445-07-2 | |
| 330.60 | |
| WSVDSBZMYJJMSB-UHFFFAOYSA-N | |
| 78187 | |
| CCCCCCCCCCCCCCCCCCC1=CC=CC=C1 |
| C24H42 | |
| MFCD00048500 | |
| 1-phenyloctadecane, benzene, octadecyl, n-octadecylbenzene, stearylbenzene, 2-phenyloctadecane, octadecane, 1-phenyl, n-octadecyl benzene, acmc-2097fm, octadecane, 1-phenyl-8ci, 1-phenyloctadecane, analytical standard | |
| octadecylbenzene |
Specifications
| 4445-07-2 | |
| C24H42 | |
| 5 g | |
| WSVDSBZMYJJMSB-UHFFFAOYSA-N | |
| octadecylbenzene | |
| 78187 | |
| ≥95.0% (GC) | |
| Octadecylbenzene |
| White-Yellow | |
| MFCD00048500 | |
| 1-phenyloctadecane, benzene, octadecyl, n-octadecylbenzene, stearylbenzene, 2-phenyloctadecane, octadecane, 1-phenyl, n-octadecyl benzene, acmc-2097fm, octadecane, 1-phenyl-8ci, 1-phenyloctadecane, analytical standard | |
| CCCCCCCCCCCCCCCCCCC1=CC=CC=C1 | |
| 330.60 | |
| 330.60 | |
| Crystalline Lumps |
Safety and Handling
EINECSNumber : (3)-0021
TSCA : Yes