missing translation for 'onlineSavingsMsg'
Learn More

Octadecylbenzene, 98%

CAS: 4445-07-2 | C24H42 | 330.59 g/mol

$269.53 - $2524.82

Chemical Identifiers

CAS 4445-07-2
Molecular Formula C24H42
Molecular Weight (g/mol) 330.59
MDL Number MFCD00048500
InChI Key WSVDSBZMYJJMSB-UHFFFAOYSA-N
IUPAC Name octadecylbenzene
SMILES CCCCCCCCCCCCCCCCCCC1=CC=CC=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC124700010
SDS View Documents
Thermo Scientific Chemicals
124700010
1 g Glass bottle
Each for $269.53
Only null left
 
AC124700100
SDS View Documents
Thermo Scientific Chemicals
124700100
10 g Glass bottle
Each for $672.77
Only null left
 
AC124700500
SDS View Documents
Thermo Scientific Chemicals
124700500
50 g Glass bottle
Each for $2,524.82
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4445-07-2
330.59
WSVDSBZMYJJMSB-UHFFFAOYSA-N
CCCCCCCCCCCCCCCCCCC1=CC=CC=C1
C24H42
MFCD00048500
octadecylbenzene

Specifications

4445-07-2
100.0
White to Yellow
>112°C
98%
C24H42
MFCD00048500
1-phenyloctadecane, benzene, octadecyl, n-octadecylbenzene, stearylbenzene, 2-phenyloctadecane, octadecane, 1-phenyl, n-octadecyl benzene, acmc-2097fm, octadecane, 1-phenyl-8ci, 1-phenyloctadecane, analytical standard
CCCCCCCCCCCCCCCCCCC1=CC=CC=C1
330.59
330.59
Low Melting Solid
97.5
32°C to 36°C
249°C (15.0 mmHg)
Authentic
Glass bottle
C6H5(CH2)17CH3
1 g
WSVDSBZMYJJMSB-UHFFFAOYSA-N
octadecylbenzene
78187
98%
Octadecylbenzene
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 224-684-7

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only