missing translation for 'onlineSavingsMsg'
Learn More

Octadecanedioic Acid 98.0+%, TCI America™

Catalog No. O02221G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02221G 1 g
O02225G 5 g
O022225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. O02221G Supplier TCI America Supplier No. O02221G
Only null left

Chemical Identifiers

CAS 871-70-5
Molecular Formula C18H34O4
Molecular Weight (g/mol) 314.466
MDL Number MFCD00142369
InChI Key BNJOQKFENDDGSC-UHFFFAOYSA-N
Synonym 1,16-hexadecanedicarboxylic acid, 1,18-octadecanedioic acid, unii-rsz6pq0qqj, octadecane-1,18-dioic acid, rsz6pq0qqj, hexadecanedicarboxylic acid, 1,18-octadecadioic acid, octadecanedioate, 1,18-octadecanedioate, octadecandis
PubChem CID 70095
IUPAC Name octadecanedioic acid
SMILES C(CCCCCCCCC(=O)O)CCCCCCCC(=O)O

Specifications

Melting Point 125°C
Color White
Quantity 1 g
Formula Weight 314.47
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Octadecanedioic Acid
EINECSNumber (2)-4022
TSCA Yes