missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octadecamethylcyclononasiloxane 98.0+%, TCI America™
$905.94 - $2,712.21
Chemical Identifiers
| CAS | 556-71-8 |
|---|---|
| Molecular Formula | C18H54O9Si9 |
| Molecular Weight (g/mol) | 667.386 |
| InChI Key | ISXOGOLHEGHGQF-UHFFFAOYSA-N |
| PubChem CID | 11172 |
| IUPAC Name | 2,2,4,4,6,6,8,8,10,10,12,12,14,14,16,16,18,18-octadecamethyl-1,3,5,7,9,11,13,15,17-nonaoxa-2,4,6,8,10,12,14,16,18-nonasilacyclooctadecane |
| SMILES | C[Si]1(O[Si](O[Si](O[Si](O[Si](O[Si](O[Si](O[Si](O[Si](O1)(C)C)(C)C)(C)C)(C)C)(C)C)(C)C)(C)C)(C)C)C |
Chemical Identifiers
| 556-71-8 | |
| 667.386 | |
| 11172 | |
| C[Si]1(O[Si](O[Si](O[Si](O[Si](O[Si](O[Si](O[Si](O[Si](O1)(C)C)(C)C)(C)C)(C)C)(C)C)(C)C)(C)C)(C)C)C |
| C18H54O9Si9 | |
| ISXOGOLHEGHGQF-UHFFFAOYSA-N | |
| 2,2,4,4,6,6,8,8,10,10,12,12,14,14,16,16,18,18-octadecamethyl-1,3,5,7,9,11,13,15,17-nonaoxa-2,4,6,8,10,12,14,16,18-nonasilacyclooctadecane |
Specifications
| 556-71-8 | |
| Yellow | |
| C18H54O9Si9 | |
| ISXOGOLHEGHGQF-UHFFFAOYSA-N | |
| 2,2,4,4,6,6,8,8,10,10,12,12,14,14,16,16,18,18-octadecamethyl-1,3,5,7,9,11,13,15,17-nonaoxa-2,4,6,8,10,12,14,16,18-nonasilacyclooctadecane | |
| 11172 | |
| ≥98.0% (GC) | |
| Octadecamethylcyclononasiloxane |
| -27°C | |
| 188°C | |
| 1 mL | |
| C[Si]1(O[Si](O[Si](O[Si](O[Si](O[Si](O[Si](O[Si](O[Si](O1)(C)C)(C)C)(C)C)(C)C)(C)C)(C)C)(C)C)(C)C)C | |
| 667.386 | |
| 667.39 | |
| Liquid |
Safety and Handling
EINECSNumber : (7)-0475
TSCA : No