missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octa-O-acetyl D-(+)-Sucrose 98.0+%, TCI America™
$32.28 - $205.05
Chemical Identifiers
| CAS | 126-14-7 |
|---|---|
| Molecular Formula | C28H38O19 |
| Molecular Weight (g/mol) | 678.593 |
| MDL Number | MFCD00006623 |
| InChI Key | ZIJKGAXBCRWEOL-SAXBRCJISA-N |
| Synonym | sucrose octaacetate, octaacetylsucrose, octa-o-acetylsucrose, d-+-sucrose octaacetate, fema no. 3038, sucrose octaacetate nf, d-+-saccharose octaacetate, 2,3,4,6,1',3',4',6'-octa-o-acetylsucrose, alpha-d-glucopyranoside, 1,3,4,6-tetra-o-acetyl-beta-d-fructofuranosyl, tetraacetate |
| PubChem CID | 31340 |
| IUPAC Name | [(2R,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-[(2S,3S,4R,5R)-3,4-diacetyloxy-2,5-bis(acetyloxymethyl)oxolan-2-yl]oxyoxan-2-yl]methyl acetate |
| SMILES | CC(=O)OCC1C(C(C(C(O1)OC2(C(C(C(O2)COC(=O)C)OC(=O)C)OC(=O)C)COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C |
Chemical Identifiers
| 126-14-7 | |
| 678.593 | |
| ZIJKGAXBCRWEOL-SAXBRCJISA-N | |
| 31340 | |
| CC(=O)OCC1C(C(C(C(O1)OC2(C(C(C(O2)COC(=O)C)OC(=O)C)OC(=O)C)COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C |
| C28H38O19 | |
| MFCD00006623 | |
| sucrose octaacetate, octaacetylsucrose, octa-o-acetylsucrose, d-+-sucrose octaacetate, fema no. 3038, sucrose octaacetate nf, d-+-saccharose octaacetate, 2,3,4,6,1',3',4',6'-octa-o-acetylsucrose, alpha-d-glucopyranoside, 1,3,4,6-tetra-o-acetyl-beta-d-fructofuranosyl, tetraacetate | |
| [(2R,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-[(2S,3S,4R,5R)-3,4-diacetyloxy-2,5-bis(acetyloxymethyl)oxolan-2-yl]oxyoxan-2-yl]methyl acetate |
Specifications
| 126-14-7 | |
| White | |
| MFCD00006623 | |
| sucrose octaacetate, octaacetylsucrose, octa-o-acetylsucrose, d-+-sucrose octaacetate, fema no. 3038, sucrose octaacetate nf, d-+-saccharose octaacetate, 2,3,4,6,1',3',4',6'-octa-o-acetylsucrose, alpha-d-glucopyranoside, 1,3,4,6-tetra-o-acetyl-beta-d-fructofuranosyl, tetraacetate | |
| CC(=O)OCC1C(C(C(C(O1)OC2(C(C(C(O2)COC(=O)C)OC(=O)C)OC(=O)C)COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C | |
| 678.593 | |
| 678.59 | |
| Crystalline Powder |
| 86°C | |
| C28H38O19 | |
| 25 g | |
| ZIJKGAXBCRWEOL-SAXBRCJISA-N | |
| [(2R,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-[(2S,3S,4R,5R)-3,4-diacetyloxy-2,5-bis(acetyloxymethyl)oxolan-2-yl]oxyoxan-2-yl]methyl acetate | |
| 31340 | |
| ≥98.0% (T) | |
| Octa-O-acetyl D-(+)-Sucrose |
Safety and Handling
EINECSNumber : (8)-0067
RTECSNumber : WN6620000
TSCA : Yes