missing translation for 'onlineSavingsMsg'
Learn More

Octa-O-acetyl D-(+)-Sucrose 98.0+%, TCI America™

Catalog No. S005225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S005225G 25 g
S0052500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S005225G Supplier TCI America Supplier No. S005225G

Chemical Identifiers

CAS 126-14-7
Molecular Formula C28H38O19
Molecular Weight (g/mol) 678.593
MDL Number MFCD00006623
InChI Key ZIJKGAXBCRWEOL-SAXBRCJISA-N
Synonym sucrose octaacetate, octaacetylsucrose, octa-o-acetylsucrose, d-+-sucrose octaacetate, fema no. 3038, sucrose octaacetate nf, d-+-saccharose octaacetate, 2,3,4,6,1',3',4',6'-octa-o-acetylsucrose, alpha-d-glucopyranoside, 1,3,4,6-tetra-o-acetyl-beta-d-fructofuranosyl, tetraacetate
PubChem CID 31340
IUPAC Name [(2R,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-[(2S,3S,4R,5R)-3,4-diacetyloxy-2,5-bis(acetyloxymethyl)oxolan-2-yl]oxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)OC2(C(C(C(O2)COC(=O)C)OC(=O)C)OC(=O)C)COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Melting Point 86°C
Color White
Quantity 25 g
Formula Weight 678.59
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Octa-O-acetyl D-(+)-Sucrose
EINECSNumber (8)-0067
RTECSNumber WN6620000
TSCA Yes