missing translation for 'onlineSavingsMsg'
Learn More

Medicom™ Ocean Pacific Ninja ™ Nitrile Gloves

Catalog No. 296367492
Change view
Click to view available options
Size:
Small
Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
296367492 Small
296367493 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 296367492 Supplier Medicom™ Supplier No. MEDOPSNI
Edge

Fully textured, powder free black nitrile glove with superior resistance and grip. Puncture resistance provides enhanced protection against bloodborne pathogens, chemical lubricants and sharp instruments.

  • Thick and durable with ample flex for the fine artistry of tattoo, hair design, dentistry, ambulance, laboratory science or pet care.
  • Fully textured to ensure a secure grip, even when wet.
  • Superior puncture resistance provides enhanced protection against bloodborne pathogens, chemical lubricants and sharp instruments.
  • ASTM D6978-05 certified for chemotherapy handling.
  • Evaluated and considered acceptable by Health Canada for food handling.

Specifications

Chlorinated No
Accelerator Free No
Applications Healthcare, Industrial, Laboratory
AQL 1.5
Color Black
Disposable Single-use
Exterior Finish Fully Textured
Latex Free Yes
Powdered Powder Free
Size Small
Sterility Non-sterile
FDA 510 (k) Yes
Glove Cuff Style Beaded
Show More Show Less