missing translation for 'onlineSavingsMsg'
Learn More

New Pig Obsidian™ Lithium Battery Containment Bag Kit

Catalog No. p-200136132
Change view
Click to view available options
Capacity (English):
10000 mAh
20000 mAh
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
17987821 10000 mAh
17987822 20000 mAh
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17987821 Supplier New Pig Supplier No. BAT320
Only null left

Secure suspect lithium batteries inside this fire-resistant bag to minimize damage, reduce spread and help prevent secondary fires

  • The Obsidian™ Lithium Battery Containment Bag Kit is designed for the safe handling and transport of lithium batteries
  • The kit includes a specialized bag to contain a damaged, recalled, or defective battery
  • Holds 10000 mAh Capacity with Lithium-ion Battery Fire Isolation and Suppression
  • Kit also includes face shield, apron and gloves with 15.12 in. W x 18.75 in. L
  • It is engineered to mitigate the risks associated with thermal runaway events in lithium-ion batteries
  • The product provides a secure solution for compliance with shipping and safety regulations
  • This kit is an essential safety item for anyone handling or transporting potentially hazardous lithium batteries

Specifications

Capacity (English) 10000 mAh
Color Orange, Clear, Yellow, Green, Black
Material Polypropylene, Aramid Fiber
Product Type Lithium Battery Containment Kit