missing translation for 'onlineSavingsMsg'
Learn More
Learn More
New Pig Obsidian™ Lithium Battery Containment Bag Kit
Description
- The Obsidian™ Lithium Battery Containment Bag Kit is designed for the safe handling and transport of lithium batteries
- The kit includes a specialized bag to contain a damaged, recalled, or defective battery
- Holds 10000 mAh Capacity with Lithium-ion Battery Fire Isolation and Suppression
- Kit also includes face shield, apron and gloves with 15.12 in. W x 18.75 in. L
- It is engineered to mitigate the risks associated with thermal runaway events in lithium-ion batteries
- The product provides a secure solution for compliance with shipping and safety regulations
- This kit is an essential safety item for anyone handling or transporting potentially hazardous lithium batteries
Spécifications
Spécifications
| Capacity (English) | 10000 mAh |
| Color | Orange, Clear, Yellow, Green, Black |
| Material | Polypropylene, Aramid Fiber |
| Product Type | Lithium Battery Containment Kit |