Learn More
Waters Limited Oasis WAX Online Column, 80, 30 m, 3.9 mm X 20 mm, 1/pk
Catalog No. 50781165
Encompass
The Oasis WAX on-line column is designed for rugged, reproducible, and ultra-fast on-line SPE. This column contains the Oasis WAX sorbent, which is a polymeric reversed-phase, weak anion exchange mixed-mode sorbent that has been optimized for fast, simple, and highly selective sample preparation of strong acidic compounds. This on-line column is designed to be used with the appropriate Waters narrow-bore analytical column, such as XSelect, XBridge, SunFire, Atlantis, XTerra, or Symmetry column. The column features traditional HPLC column fittings and hardware.
Catalog No. 50781165 Supplier Waters Limited Supplier No. 186002509
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The Oasis WAX on-line column is designed for rugged, reproducible, and ultra-fast on-line SPE. This column contains the Oasis WAX sorbent, which is a polymeric reversed-phase, weak anion exchange mixed-mode sorbent that has been optimized for fast, simple, and highly selective sample preparation of strong acidic compounds. This on-line column is designed to be used with the appropriate Waters narrow-bore analytical column, such as XSelect, XBridge, SunFire, Atlantis, XTerra, or Symmetry column. The column features traditional HPLC column fittings and hardware.