Learn More
Waters Limited Oasis WAX 6 cc Vac Cartridge, 150 mg Sorbent per Cartridge, 30 m, 30/pk
Supplier:  Waters Limited 186002493
Encompass
The 6 cc Oasis WAX cartridges contain the Oasis WAX sorbent, which is a polymeric reversed-phase, weak anion exchange mixed-mode sorbent that has been optimized for fast, simple, and highly selective sample preparation of strong acidic compounds. Since the Oasis WAX sorbent is water wettable, it maintains its capability for higher retention and excellent recoveries even if the sorbent runs dry, which means there is no need to take extraordinary steps to keep the sorbent beds from drying out during the critical steps prior to sample loading.
Catalog No. 50781150
Only null left