Learn More
Cole-Parmer™ Oakton™ Dissolved Oxygen Meter Probes
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample.
Supplier:  Cole-Parmer™ 3564050
Catalog No. 15500034
Only null left
Description
Description
DO/temperature probes with submersible cables. Model with 10 ft. cable is a replacement and 25 ft. model an accessory for Oakton Dissolved Oxygen Meters DO 100, DO 110, DO 300, ph/DO 300.
Specifications
Specifications
10 ft.
DO
Oakton 600 series DO meters, Accumet AP84, AP74 and Oakton DO 100 portable meter
DO
DO/Temperature
3 m
50mL of electrolyte solution and two replacement membrane caps
17.3 cm
6.8 in.
DO, Temp
Product Suggestions
Product Suggestions