Learn More
Oakton™ Water Analysis Accessories
For your convenience, cables, power adapters, and other accessories are available for your Oakton meters and pocket testers.
Supplier: Oakton™ 3564075
Description
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sampleSpecifications
| Oakton Replacement DO Probe Membranes; 25/Pk | |
| Membrane | |
| 25 membranes - membrane replacement tool (35640-79) required; sold separately | |
| DO |
| Oakton Meters: DO 6, DO 100, DO 110, DO 300 | |
| DO measurement | |
| HDPE |