missing translation for 'onlineSavingsMsg'
Learn More

o-Xylylene Oxide 97.0+%, TCI America™

Catalog No. p-7137966
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X00535G 5 g
X005325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. X00535G Supplier TCI America Supplier No. X00535G
Only null left

Chemical Identifiers

CAS 496-14-0
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.151
MDL Number MFCD00005932
InChI Key SFLGSKRGOWRGBR-UHFFFAOYSA-N
Synonym phthalan, 1,3-dihydroisobenzofuran, isocoumaran, isobenzofuran, 1,3-dihydro, o-xylylene oxide, 2-oxaindan, 1,3-dihydro isobenzofuran, unii-2r6ntq7y6g, ccris 1557, 2r6ntq7y6g
PubChem CID 10327
IUPAC Name 1,3-dihydro-2-benzofuran
SMILES C1C2=CC=CC=C2CO1

Specifications

Color Colorless
Boiling Point 192°C
Quantity 5 g
Formula Weight 120.15
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material o-Xylylene Oxide