missing translation for 'onlineSavingsMsg'
Learn More

o-Tolylacetic Acid 98.0+%, TCI America™

Catalog No. p-7136478
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T088125G 25 g
T0881250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T088125G Supplier TCI America Supplier No. T088125G

Chemical Identifiers

CAS 644-36-0
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00004328
InChI Key RZWGTXHSYZGXKF-UHFFFAOYSA-N
Synonym o-tolylacetic acid, 2-methylphenylacetic acid, 2-methylphenyl acetic acid, 2-2-methylphenyl acetic acid, 2-o-tolyl acetic acid, 2-tolylacetic acid, 2-o-tolylacetic acid, o-methylphenylacetic acid, o-tolyacetic acid
PubChem CID 69519
IUPAC Name 2-(2-methylphenyl)acetic acid
SMILES CC1=CC=CC=C1CC(=O)O

Specifications

Melting Point 89°C
Color White
Quantity 25 g
Formula Weight 150.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material o-Tolylacetic Acid