missing translation for 'onlineSavingsMsg'
Learn More

o-Tolylacetic acid, 99%

Catalog No. AC139100250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC139100250 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC139100250 Supplier Thermo Scientific Chemicals Supplier No. 139100250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 644-36-0 | C9H10O2 | 150.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 644-36-0
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00004328
InChI Key RZWGTXHSYZGXKF-UHFFFAOYSA-N
Synonym o-tolylacetic acid, 2-methylphenylacetic acid, 2-methylphenyl acetic acid, 2-2-methylphenyl acetic acid, 2-o-tolyl acetic acid, 2-tolylacetic acid, 2-o-tolylacetic acid, o-methylphenylacetic acid, o-tolyacetic acid
PubChem CID 69519
IUPAC Name 2-(2-methylphenyl)acetic acid
SMILES CC1=CC=CC=C1CC(=O)O

Specifications

Melting Point 87°C to 90°C
Color Cream to White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula CH3C6H4CH2CO2H
Quantity 25 g
Beilstein 09, 527
Formula Weight 150.18
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material o-Tolylacetic acid, 99%
Show More Show Less
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only