missing translation for 'onlineSavingsMsg'
Learn More

o-Tolylacetic acid, 99%

Catalog No. AAA1578306
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1578306 5 g
AAA1578314 25 g
AAA1578322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1578306 Supplier Thermo Scientific Chemicals Supplier No. A1578306

CAS: 644-36-0 | C9H10O2 | 150.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 644-36-0
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00004328
InChI Key RZWGTXHSYZGXKF-UHFFFAOYSA-N
Synonym o-tolylacetic acid, 2-methylphenylacetic acid, 2-methylphenyl acetic acid, 2-2-methylphenyl acetic acid, 2-o-tolyl acetic acid, 2-tolylacetic acid, 2-o-tolylacetic acid, o-methylphenylacetic acid, o-tolyacetic acid
PubChem CID 69519
IUPAC Name 2-(2-methylphenyl)acetic acid
SMILES CC1=CC=CC=C1CC(=O)O

Specifications

Melting Point 86°C to 90°C
Quantity 5 g
Beilstein 1936952
Formula Weight 150.18
Percent Purity 99%
Chemical Name or Material o-Tolylacetic acid
Hazard Category H315-H319-H335-H500
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-416-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only