missing translation for 'onlineSavingsMsg'
Learn More

o-Tolyl isocyanate, 98%

Catalog No. p-7045858
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1224706 5 g
AAL1224714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1224706 Supplier Thermo Scientific Chemicals Supplier No. L1224706
Only null left

CAS: 614-68-6 | C8H7NO | 133.15 g/mol

o-Tolyl isocyanate is used as plastic additives and as heat treatment salt formulations for metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-Tolyl isocyanate is used as plastic additives and as heat treatment salt formulations for metals.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Incompatible with bases, moisture and oxidizing agents. Store under dry inert gas. Protect from humidity and water.

Chemical Identifiers

CAS 614-68-6
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00002009
InChI Key VAYMIYBJLRRIFR-UHFFFAOYSA-N
Synonym o-tolyl isocyanate, 2-methylphenyl isocyanate, 2-tolyl isocyanate, o-methylphenyl isocyanate, o-toluene isocyanate, benzene, 1-isocyanato-2-methyl, 2-methylphenylisocyanate, tolyl isocyanate, isocyanic acid, o-tolyl ester, o-tolylisocyanate
PubChem CID 69194
ChEBI CHEBI:60099
IUPAC Name 1-isocyanato-2-methylbenzene
SMILES CC1=CC=CC=C1N=C=O

Specifications

Density 1.074
Boiling Point 186°C
Flash Point 92°C (197°F)
Odor Pungent
Refractive Index 1.5345
Quantity 5 g
UN Number UN2206
Beilstein 507985
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 133.15
Percent Purity 98%
Chemical Name or Material o-Tolyl isocyanate
Show More Show Less
Hazard Category H227-H302-H314-H332-H334-H335
Hazard Statement GHS H Statement
H330-H334-H227-H302-H315-H319-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 210-389-0
RTECSNumber DA3678333
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only