missing translation for 'onlineSavingsMsg'
Learn More

o-Tolualdehyde 2,4-Dinitrophenylhydrazone 98.0+%, TCI America™

Catalog No. p-7136781
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14471G 1 g
T144710G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T14471G Supplier TCI America Supplier No. T14471G
Only null left

Chemical Identifiers

CAS 1773-44-0
Molecular Formula C14H12N4O4
Molecular Weight (g/mol) 300.27
MDL Number MFCD00191584
InChI Key QUIFNJPMGQCNMH-OQLLNIDSSA-N
PubChem CID 9555686
IUPAC Name (2E)-1-(2,4-dinitrophenyl)-2-[(2-methylphenyl)methylidene]hydrazine
SMILES CC1=CC=CC=C1\C=N\NC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O

Specifications

Melting Point 196°C
Quantity 1 g
UN Number 1325
Synonym 2′C-Methylbenzaldehyde 2,4-Dinitrophenylhydrazone
Formula Weight 300.27
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material o-Tolualdehyde 2,4-Dinitrophenylhydrazone