Learn More
O-(tert-Butyldimethylsilyl)hydroxylamine, 90+%, Thermo Scientific Chemicals
Description
O-(tert-Butyldimethylsilyl)hydroxylamine (tert-butyldimethylsilyl-O-hydroxylamine)is used to induce the cleavage, during solid supported synthesis of hydroxamic acids. It also finds it uses as a chemical additive and an intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
O-(tert-Butyldimethylsilyl)hydroxylamine (tert-butyldimethylsilyl-O-hydroxylamine)is used to induce the cleavage, during solid supported synthesis of hydroxamic acids. It also finds it uses as a chemical additive and an intermediate.
Solubility
Hydrolyzes in water.
Notes
Moisture sensitive. Store away from oxidizing agents, water/moisture and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications
Specifications
| Boiling Point | 87°C to 90°C (40mmHg) |
| Melting Point | 62°C to 65°C |
| Quantity | 1 g |
| UN Number | UN3259 |
| Solubility Information | Hydrolyzes in water. |
| Formula Weight | 147.29 |
| Percent Purity | ≥90% |
| Sensitivity | Moisture sensitive |
| Chemical Name or Material | O-(tert-Butyldimethylsilyl)hydroxylamine |
Safety and Handling
| Hazard Category | H314-H335 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S. |
| TSCA | No |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only