missing translation for 'onlineSavingsMsg'
Learn More

O-(tert-Butyldimethylsilyl)hydroxylamine, 90+%, Thermo Scientific Chemicals

Catalog No. AAB2100103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2100103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2100103 Supplier Thermo Scientific Chemicals Supplier No. B2100103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 41879-39-4 | Molecular Formula: C6H17NOSi | Molecular Weight: 147.293 g/mol

O-(tert-Butyldimethylsilyl)hydroxylamine (tert-butyldimethylsilyl-O-hydroxylamine)is used to induce the cleavage, during solid supported synthesis of hydroxamic acids. It also finds it uses as a chemical additive and an intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-(tert-Butyldimethylsilyl)hydroxylamine (tert-butyldimethylsilyl-O-hydroxylamine)is used to induce the cleavage, during solid supported synthesis of hydroxamic acids. It also finds it uses as a chemical additive and an intermediate.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store away from oxidizing agents, water/moisture and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Chemical Identifiers

CAS 41879-39-4
Molecular Formula C6H17NOSi
Molecular Weight (g/mol) 147.293
MDL Number MFCD00054944
InChI Key SSUCKKNRCOFUPT-UHFFFAOYSA-N
Synonym o-tert-butyldimethylsilyl hydroxylamine, o-t-butyldimethylsilyl hydroxylamine, aminoxy-tert-butyldimethylsilane, o-tert-butyldimethylsilylhydroxylamine, o-tert-butyl dimethyl silyl hydroxylamine, h2n-otbs, tbs-onh2, acmc-1asp9, hydroxylamine, tbdms derivative, o-t-butyldimethylsilylhydroxylamine
PubChem CID 4691350
IUPAC Name O-[tert-butyl(dimethyl)silyl]hydroxylamine
SMILES CC(C)(C)[Si](C)(C)ON

Specifications

Boiling Point 87°C to 90°C (40mmHg)
Melting Point 62°C to 65°C
Quantity 1 g
UN Number UN3259
Solubility Information Hydrolyzes in water.
Formula Weight 147.29
Percent Purity ≥90%
Sensitivity Moisture sensitive
Chemical Name or Material O-(tert-Butyldimethylsilyl)hydroxylamine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only