missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-tert-Butyl-N,N'-diisopropylisourea 98.0+%, TCI America™
$139.81 - $475.33
Chemical Identifiers
| CAS | 71432-55-8 |
|---|---|
| Molecular Formula | C11H24N2O |
| Molecular Weight (g/mol) | 200.326 |
| MDL Number | MFCD06657672 |
| InChI Key | FESDUDPSRMWIDL-UHFFFAOYSA-N |
| Synonym | 2-tert-Butyl-1,3-diisopropylisourea |
| PubChem CID | 11041782 |
| IUPAC Name | tert-butyl N,N'-di(propan-2-yl)carbamimidate |
| SMILES | CC(C)NC(=NC(C)C)OC(C)(C)C |
Chemical Identifiers
| 71432-55-8 | |
| 200.326 | |
| FESDUDPSRMWIDL-UHFFFAOYSA-N | |
| 11041782 | |
| CC(C)NC(=NC(C)C)OC(C)(C)C |
| C11H24N2O | |
| MFCD06657672 | |
| 2-tert-Butyl-1,3-diisopropylisourea | |
| tert-butyl N,N'-di(propan-2-yl)carbamimidate |
Specifications
| 71432-55-8 | |
| 61°C | |
| MFCD06657672 | |
| 2-tert-Butyl-1,3-diisopropylisourea | |
| CC(C)NC(=NC(C)C)OC(C)(C)C | |
| 200.326 | |
| 200.33 | |
| Liquid |
| Colorless | |
| C11H24N2O | |
| 1 g | |
| FESDUDPSRMWIDL-UHFFFAOYSA-N | |
| tert-butyl N,N'-di(propan-2-yl)carbamimidate | |
| 11041782 | |
| ≥98.0% (T) | |
| O-tert-Butyl-N,N′-diisopropylisourea |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator