missing translation for 'onlineSavingsMsg'
Learn More

O-tert-Butyl-N,N'-diisopropylisourea 98.0+%, TCI America™

Catalog No. B41781G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41781G 1 g
B41785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41781G Supplier TCI America Supplier No. B41781G
Only null left

Chemical Identifiers

CAS 71432-55-8
Molecular Formula C11H24N2O
Molecular Weight (g/mol) 200.326
MDL Number MFCD06657672
InChI Key FESDUDPSRMWIDL-UHFFFAOYSA-N
Synonym 2-tert-Butyl-1,3-diisopropylisourea
PubChem CID 11041782
IUPAC Name tert-butyl N,N'-di(propan-2-yl)carbamimidate
SMILES CC(C)NC(=NC(C)C)OC(C)(C)C

Specifications

Color Colorless
Boiling Point 61°C
Quantity 1 g
Formula Weight 200.33
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material O-tert-Butyl-N,N′-diisopropylisourea
TSCA No
Recommended Storage Refrigerator