missing translation for 'onlineSavingsMsg'
Learn More

O-tert-Butyl-L-threonine, 97%

CAS: 4378-13-6 | C8H17NO3 | 175.23 g/mol

$136.81 - $488.99

Chemical Identifiers

CAS 4378-13-6
Molecular Formula C8H17NO3
Molecular Weight (g/mol) 175.23
MDL Number MFCD00037766
InChI Key NMJINEMBBQVPGY-RITPCOANSA-N
Synonym h-thr tbu-oh, o-tert-butyl-l-threonine, 2s,3r-2-amino-3-tert-butoxy butanoic acid, l-threonine, o-1,1-dimethylethyl, o-t-butyl-l-threonine, 2s,3r-2-amino-3-2-methylpropan-2-yl oxy butanoic acid, o-tert.-butylthreonin, o-tert.butyl-l-threonine, h-ser trt-2-chlorotrityl resin, h-thr tbu-2-chlorotrityl resin
PubChem CID 6951357
IUPAC Name (2S,3R)-2-amino-3-[(2-methylpropan-2-yl)oxy]butanoic acid
SMILES C[C@@H](OC(C)(C)C)[C@H](N)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6303106
View Documents
Thermo Scientific Chemicals
H6303106
5 g
Each for $136.81
Only null left
 
AAH6303114
View Documents
Thermo Scientific Chemicals
H6303114
25 g
Each for $488.99
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4378-13-6
175.23
NMJINEMBBQVPGY-RITPCOANSA-N
6951357
C[C@@H](OC(C)(C)C)[C@H](N)C(O)=O
C8H17NO3
MFCD00037766
h-thr tbu-oh, o-tert-butyl-l-threonine, 2s,3r-2-amino-3-tert-butoxy butanoic acid, l-threonine, o-1,1-dimethylethyl, o-t-butyl-l-threonine, 2s,3r-2-amino-3-2-methylpropan-2-yl oxy butanoic acid, o-tert.-butylthreonin, o-tert.butyl-l-threonine, h-ser trt-2-chlorotrityl resin, h-thr tbu-2-chlorotrityl resin
(2S,3R)-2-amino-3-[(2-methylpropan-2-yl)oxy]butanoic acid

Specifications

4378-13-6
MFCD00037766
h-thr tbu-oh, o-tert-butyl-l-threonine, 2s,3r-2-amino-3-tert-butoxy butanoic acid, l-threonine, o-1,1-dimethylethyl, o-t-butyl-l-threonine, 2s,3r-2-amino-3-2-methylpropan-2-yl oxy butanoic acid, o-tert.-butylthreonin, o-tert.butyl-l-threonine, h-ser trt-2-chlorotrityl resin, h-thr tbu-2-chlorotrityl resin
C[C@@H](OC(C)(C)C)[C@H](N)C(O)=O
5 g
6951357
97%
O-tert-Butyl-L-threonine
C8H17NO3
2206127
NMJINEMBBQVPGY-RITPCOANSA-N
(2S,3R)-2-amino-3-[(2-methylpropan-2-yl)oxy]butanoic acid
175.23
175.23
Solid
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only