missing translation for 'onlineSavingsMsg'
Learn More

O-tert-Butyl-L-threonine, 97%

Catalog No. AAH6303106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6303106 5 g
AAH6303114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6303106 Supplier Thermo Scientific Chemicals Supplier No. H6303106
Only null left

CAS: 4378-13-6 | C8H17NO3 | 175.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4378-13-6
Molecular Formula C8H17NO3
Molecular Weight (g/mol) 175.23
MDL Number MFCD00037766
InChI Key NMJINEMBBQVPGY-RITPCOANSA-N
Synonym h-thr tbu-oh, o-tert-butyl-l-threonine, 2s,3r-2-amino-3-tert-butoxy butanoic acid, l-threonine, o-1,1-dimethylethyl, o-t-butyl-l-threonine, 2s,3r-2-amino-3-2-methylpropan-2-yl oxy butanoic acid, o-tert.-butylthreonin, o-tert.butyl-l-threonine, h-ser trt-2-chlorotrityl resin, h-thr tbu-2-chlorotrityl resin
PubChem CID 6951357
IUPAC Name (2S,3R)-2-amino-3-[(2-methylpropan-2-yl)oxy]butanoic acid
SMILES C[C@@H](OC(C)(C)C)[C@H](N)C(O)=O

Specifications

Beilstein 2206127
Quantity 5 g
Formula Weight 175.23
Percent Purity 97%
Physical Form Solid
Chemical Name or Material O-tert-Butyl-L-threonine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only