missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-tert-Butyl-L-serine methyl ester hydrochloride, 98%
CAS: 17114-97-5 | C8H18ClNO3 | 211.69 g/mol
$54.33 - $537.96
Chemical Identifiers
| CAS | 17114-97-5 |
|---|---|
| Molecular Formula | C8H18ClNO3 |
| Molecular Weight (g/mol) | 211.69 |
| MDL Number | MFCD00077108 |
| InChI Key | PCIABNBULSRKSU-UHFFFAOYNA-N |
| Synonym | h-ser tbu-ome.hcl, o-tert-butyl-l-serine methyl ester hydrochloride, h-ser tbu-ome hcl, s-methyl 2-amino-3-tert-butoxy propanoate hydrochloride, methyl o-tert-butyl-l-serinate hydrochloride, l-serine, o-1,1-dimethylethyl-, methyl ester, hydrochloride, methyl 2s-2-amino-3-tert-butoxy propanoate hydrochloride, ksc491o3r, h-ser tbu-och3 hydrochloride, s-methyl 2-amino-3-tert-butoxy propanoate hcl |
| PubChem CID | 16218549 |
| SMILES | Cl.COC(=O)C(N)COC(C)(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAH62126MD
|
N/A | 250 mg |
Each for $54.33
|
|
|||||
|
AAH6212603
|
N/A | 1 g |
Each for $139.44
|
|
|||||
|
AAH6212606
|
N/A | 5 g |
Each for $537.96
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 17114-97-5 | |
| 211.69 | |
| PCIABNBULSRKSU-UHFFFAOYNA-N | |
| 16218549 |
| C8H18ClNO3 | |
| MFCD00077108 | |
| h-ser tbu-ome.hcl, o-tert-butyl-l-serine methyl ester hydrochloride, h-ser tbu-ome hcl, s-methyl 2-amino-3-tert-butoxy propanoate hydrochloride, methyl o-tert-butyl-l-serinate hydrochloride, l-serine, o-1,1-dimethylethyl-, methyl ester, hydrochloride, methyl 2s-2-amino-3-tert-butoxy propanoate hydrochloride, ksc491o3r, h-ser tbu-och3 hydrochloride, s-methyl 2-amino-3-tert-butoxy propanoate hcl | |
| Cl.COC(=O)C(N)COC(C)(C)C |
Specifications
| 17114-97-5 | |
| MFCD00077108 | |
| h-ser tbu-ome.hcl, o-tert-butyl-l-serine methyl ester hydrochloride, h-ser tbu-ome hcl, s-methyl 2-amino-3-tert-butoxy propanoate hydrochloride, methyl o-tert-butyl-l-serinate hydrochloride, l-serine, o-1,1-dimethylethyl-, methyl ester, hydrochloride, methyl 2s-2-amino-3-tert-butoxy propanoate hydrochloride, ksc491o3r, h-ser tbu-och3 hydrochloride, s-methyl 2-amino-3-tert-butoxy propanoate hcl | |
| Cl.COC(=O)C(N)COC(C)(C)C | |
| 211.69 | |
| 211.69 | |
| O-tert-Butyl-L-serine methyl ester hydrochloride |
| C8H18ClNO3 | |
| 2041979 | |
| PCIABNBULSRKSU-UHFFFAOYNA-N | |
| methyl 2-amino-3-(tert-butoxy)propanoate hydrochloride | |
| 16218549 | |
| 98% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only