missing translation for 'onlineSavingsMsg'
Learn More

O-(tert-Butyl)hydroxylamine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7123632
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
B45891G 1 g
B45895G 5 g
2 options

Catalog No. B45891G

Supplier: TCI America B45891G

Only null left

Chemical Identifiers

CAS 39684-28-1
Molecular Formula C4H12ClNO
Molecular Weight (g/mol) 125.596
MDL Number MFCD00043272
InChI Key ZBDXGNXNXXPKJI-UHFFFAOYSA-N
Synonym o-tert-butyl hydroxylamine hydrochloride, o-tert-butylhydroxylamine hydrochloride, o-t-butylhydroxylamine hydrochloride, tert-butoxyamine hydrochloride, o-tert-butyl hydroxylaminehydrochloride, hydroxylamine, o-1,1-dimethylethyl-, hydrochloride, soyheadhmuup@, o-tert-butylhydroxylamine hcl, pubchem19621, t-butoxyamine hydrochloride
PubChem CID 2777906
IUPAC Name O-tert-butylhydroxylamine;hydrochloride
SMILES CC(C)(C)ON.Cl

Specifications

Melting Point 156°C
Color White-Yellow
Quantity 1 g
Formula Weight 125.60
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material O-(tert-Butyl)hydroxylamine Hydrochloride