missing translation for 'onlineSavingsMsg'
Learn More

O-(tert-Butyl)hydroxylamine hydrochloride, 99%

Catalog No. AC399680050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC399680050 5 g Glass bottle
AC399680010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC399680050 Supplier Thermo Scientific Chemicals Supplier No. 399680050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 39684-28-1 | C4H11NO·HCl | 125.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39684-28-1
Molecular Formula C4H11NO·HCl
Molecular Weight (g/mol) 125.6
InChI Key ZBDXGNXNXXPKJI-UHFFFAOYSA-N
Synonym o-tert-butyl hydroxylamine hydrochloride, o-tert-butylhydroxylamine hydrochloride, o-t-butylhydroxylamine hydrochloride, tert-butoxyamine hydrochloride, o-tert-butyl hydroxylaminehydrochloride, hydroxylamine, o-1,1-dimethylethyl-, hydrochloride, soyheadhmuup@, o-tert-butylhydroxylamine hcl, pubchem19621, t-butoxyamine hydrochloride
PubChem CID 2777906
IUPAC Name O-tert-butylhydroxylamine;hydrochloride
SMILES CC(C)(C)ON.Cl

Specifications

Melting Point 155.0°C
Assay Percent Range 98.5 to 101.5% (Argentometry)
Packaging Glass bottle
Linear Formula (CH3)3CONH2·HCl
Quantity 5 g
Formula Weight 125.6
Percent Purity 99%
Chemical Name or Material O-(tert-Butyl)hydroxylamine hydrochloride
Hazard Category GHS Signal Word: Warning
EINECSNumber 254-590-1

RUO – Research Use Only