missing translation for 'onlineSavingsMsg'
Learn More
Learn More
o-Terphenyl 99.0+%, TCI America™
$125.13 - $1183.98
Chemical Identifiers
| CAS | 84-15-1 |
|---|---|
| Molecular Formula | C18H14 |
| Molecular Weight (g/mol) | 230.31 |
| MDL Number | MFCD00003055 |
| InChI Key | OIAQMFOKAXHPNH-UHFFFAOYSA-N |
| Synonym | o-terphenyl, 1,1':2',1-terphenyl, terphenyl, chlorinated, 1,1'-biphenyl, 2-phenyl, caswell no. 672b, o-diphenylbenzene, polychlorinated terphenyl, polychlorinated terphenyls, unii-w5675r7kvw, ccris 1655 |
| PubChem CID | 6766 |
| IUPAC Name | 1,2-diphenylbenzene |
| SMILES | C1=CC=C(C=C1)C2=CC=CC=C2C3=CC=CC=C3 |
Chemical Identifiers
| 84-15-1 | |
| 230.31 | |
| OIAQMFOKAXHPNH-UHFFFAOYSA-N | |
| 6766 | |
| C1=CC=C(C=C1)C2=CC=CC=C2C3=CC=CC=C3 |
| C18H14 | |
| MFCD00003055 | |
| o-terphenyl, 1,1':2',1-terphenyl, terphenyl, chlorinated, 1,1'-biphenyl, 2-phenyl, caswell no. 672b, o-diphenylbenzene, polychlorinated terphenyl, polychlorinated terphenyls, unii-w5675r7kvw, ccris 1655 | |
| 1,2-diphenylbenzene |
Specifications
| 84-15-1 | |
| White | |
| C18H14 | |
| 25 g | |
| o-terphenyl, 1,1':2',1-terphenyl, terphenyl, chlorinated, 1,1'-biphenyl, 2-phenyl, caswell no. 672b, o-diphenylbenzene, polychlorinated terphenyl, polychlorinated terphenyls, unii-w5675r7kvw, ccris 1655 | |
| C1=CC=C(C=C1)C2=CC=CC=C2C3=CC=CC=C3 | |
| 230.31 | |
| 230.31 | |
| Crystalline Powder |
| 58°C | |
| 332°C | |
| MFCD00003055 | |
| 3077 | |
| OIAQMFOKAXHPNH-UHFFFAOYSA-N | |
| 1,2-diphenylbenzene | |
| 6766 | |
| ≥99.0% (GC) | |
| o-Terphenyl |
Safety and Handling
EINECSNumber : (4)-0017
RTECSNumber : WZ6472000
TSCA : Yes