missing translation for 'onlineSavingsMsg'
Learn More

O,S-Diethyl methylphosphonothioate, 97%, Thermo Scientific Chemicals

Catalog No. p-7059357
Change view
Click to view available options
Quantity:
0.5 g
2 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4446502 0.5 g
AA4446504 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4446502 Supplier Thermo Scientific Chemicals Supplier No. 04446502
Only null left

O,S-Diethyl methylphosphonothioate is used as an inhibitor of cholinesterase. It is also used in pharmaceuticals, intermediates and fine chemicals. It also serves as phosphorylating and phosphitylating agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2511-10-6
MDL Number MFCD01705977

Specifications

Density 1.094 g/mL
Boiling Point 95°C to 97°C (10 mmHg)
Molecular Formula C5H13O2PS
Refractive Index 1.474
Quantity 0.5 g
UN Number UN3278
Synonym o,s-diethyl methylphosphonothioate, o,s-diethyl methylthiophosphonate, methylphosphonothioic acid o,s-diethyl ester, osdmp, phosphonothioic acid, methyl-, o,s-diethyl ester, phosphonothioic acid, p-methyl-, o,s-diethyl ester, o-ethyl s-ethyl methylphosphonothioate, ethyl ethylsulfanyl methyl phosphinate, acmc-20ak0l, ethoxy ethylthio methyl phosphine oxide
Solubility Information Immiscible with water.
InChI Key AIMOGGQLCCAPQJ-UHFFFAOYNA-N
SMILES CCOP(=O)(C)SCC
IUPAC Name 1-[ethylsulfanyl(methyl)phosphoryl]oxyethane
Molecular Weight (g/mol) 168.19
PubChem CID 520177
Formula Weight 168.19
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material O,S-Diethyl methylphosphonothioate
Show More Show Less
Hazard Category H300+H310+H330
Hazard Statement GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Precautionary Statement P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, LIQUID, TOXIC, N.O.S.
RTECSNumber TB1085000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only